t-Boc-N-Amido-PEG7-Amine ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: O-(2-Aminoethyl)-O′-[2-(Boc-amino)ethyl]hexaethylene glycol, 25-Amino-5,8,11,14,17,20,23-heptaoxa-2-azapentacosanoic acid 1,1-dimethylethyl ester, tert-Butyl (23-amino-3,6,9,12,15,18,21-heptaoxatricosyl)carbamate, BocNH-PEG7-CH2CH2NH2, Boc-PEG-amine (n=7), Boc-NH-PEG7-NH2

A122203-100MG A122203-250MG A122203-500MG
ALA122203100MGEA 140.51 USD
ALA122203100MG ALA122203250MG ALA122203500MG
t-Boc-N-Amido-PEG7-Amine ≥98%
t-Boc-N-Amido-PEG7-Amine

Product Introduction:Our highly purified mono-protected homobifunctional polyethylene glycols display oligomer purities of more than 90%. This extremely narrow distribution of oligomers ensures homogeneous products in many applications such as conjugation reactions or introduction of solubilizing units or spacers.

Formula: C₂₁H₄₄N₂O₉
Storage Temperature: Freezer
MDL Number: MFCD03453229
CAS Number: 206265-98-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9180db514daad33d1267eb8822a610b5d1d8963f&vid=9180db514daad33d1267eb8822a610b5d1d8963f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR