Amino-PEG4-acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Amino-PEG4-acid, Amino-PEG-acid, α-Amine-ω-propionic acid tetraethylene glycol, 3-[2-[2-[2-(2-Aminoethoxy)ethoxy]ethoxy]ethoxy]propionic acid, NH2-PEG4-COOH, Carboxy-PEG4-amine, Tetraethylene glycol amino propionic acid, NH2-PEG4-CH2CH2COOH, CA(PEG)4, (PEO)4-Aminopropionic acid, 1-Amino-3,6,9,12-tetraoxapentadecan-15-oic acid, Tetraethylene glycol α-amino-ω-propionic acid

A122170-100MG A122170-1G A122170-500MG A122170-5G
ALA122170100MGEA 74.96 USD
ALA122170100MG ALA122170-1G ALA122170500MG ALA122170-5G
Amino-PEG4-acid ≥98%
Amino-PEG4-acid

Amino-dPEG4-acid has a primary amine and propionic acid terminating opposite ends of a 16-atoms long (18.0 Å) polyethylene glycol (PEG) spacer. The single molecular weight PEG spacer is discrete (Ð = 1). Moreover, it is highly hydrophilic, and it imparts hydrophilicity to conjugates that incorporate it. Amino-dPEG4-acid is useful in many different applications, including peptide synthesis, surface modification, dendrimer construction, and small molecule modification

Formula: C₁₁H₂₃NO₆
Storage Temperature: Freezer
MDL Number: MFCD11041129
CAS Number: 663921-15-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d606821803e0d542a38b00f0c097401db9dcdacf&vid=d606821803e0d542a38b00f0c097401db9dcdacf&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR