2-Amino-3-chloropyrazine ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-Chloropyrazin-2-amine, 3-Chloro-2-pyrazinamine

A121861-1G A121861-25G A121861-5G
ALA121861-1GEA 19.86 USD
ALA121861-1G ALA121861-25G ALA121861-5G
2-Amino-3-chloropyrazine ≥97%
2-Amino-3-chloropyrazine

Product Application:It is employed in the synthesis and tuberculostatic activity of N-pyrazinylthiourea.

Formula: C₄H₄ClN₃
MW: 129.55 g/mol
Melting Pt: 168°C
MDL Number: MFCD04114305
CAS Number: 6863-73-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=584b7a39a6c4aa5a1002a4c7a3f2c56d1e998148&vid=584b7a39a6c4aa5a1002a4c7a3f2c56d1e998148&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR