3-Amino-5-hydroxy-1H-pyrazole 98

Supplier: ALADDIN SCIENTIFIC

Synonyms: 3-Amino-5-hydroxy-1H-pyrazole, 3-Amino-5-hydroxypyrazole, 5-Amino-3-pyrazolol, 3-Amino-1H-pyrazol-5(4H)-one, 3-Amino-2-pyrazolin-5-one, 5-Hydroxy-3-aminopyrazole, 5-Amino-2,4-dihydro-3H-pyrazol-3-one

A121779-100G A121779-25G A121779-5G
ALA121779-100GEA 190.86 USD
ALA121779-100G ALA121779-25G ALA121779-5G
3-Amino-5-hydroxy-1H-pyrazole 98
3-Amino-5-hydroxy-1H-pyrazole

Usually used in the synthesis of 2,6-difluoro-N-(3-methoxy-1H-pyrazolo[3,4-b]pyridine-5-yl)-3-(propylsulfonamidio)benzamide and heteroaromatic 3-hydroxy-5,6-diphenylpyrazolo[3,4-b]pyridines.

Formula: C₃H₅N₃O
MW: 99.09 g/mol
Melting Pt: 210°C
MDL Number: MFCD00022384
CAS Number: 6126-22-3
EINECS: 228-095-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR