6-Amino-m-cresol ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 1-Amino-2-hydroxy-4-methylbenzene, 2-Hydroxy-4-methylaniline, Cetilistat Impurity 4, 4-Amino-3-hydroxytoluene, 2-Amino-5-methylphenol

A121774-100G A121774-25G A121774-5G
ALA121774-100GEA 438.81 USD
ALA121774-100G ALA121774-25G ALA121774-5G
6-Amino-m-cresol ≥98%
6-Amino-m-cresol

Synthesis of tridentate Schiff base ligand and novel non-metallocene catalysts with phenoxy-imine ligands.

Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 259
Melting Pt: 162
Storage Temperature: Ambient
MDL Number: MFCD00007693
CAS Number: 2835-98-5
EINECS: 220-620-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64bf21a538812d002997c2c1e1b10cbb82b8f7b0&vid=64bf21a538812d002997c2c1e1b10cbb82b8f7b0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR