Acridine orange ≥75%

Supplier: ALADDIN SCIENTIFIC

Synonyms: Brilliant Acridine Orange E, 3,6-Bis(dimethylamino)acridine, Acridine Orange base, Solvent Orange 15, 3,6-Bis(dimethylamino)acridine N,N,N',N'-Tetramethyl-3,6-acridinediamine, Acridine orange free base, Acridine Orange Stain

A121748-1G A121748-25G A121748-5G
ALA121748-1GEA 155.71 USD
ALA121748-1G ALA121748-25G ALA121748-5G
Acridine orange ≥75%
Acridine orange

When bound to DNA, it emits green fluorescence (Em=525 nm) and when bound to RNA, it emits red fluorescence (Em=~650 nm).A fluorescent nucleic acid binding dye which interacts with both DNA and RNA.

Formula: C₁₇H₁₉N₃
MW: 265.35 g/mol
Melting Pt: 165°C
MDL Number: MFCD00005029
CAS Number: 494-38-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96d18d805db4e5d8ffb1eae19d7e08614d9d7cea&vid=96d18d805db4e5d8ffb1eae19d7e08614d9d7cea&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR