1-(2-Aminoethyl)pyrrolidine ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: N-(2-Aminoethyl)pyrrolidine, Pyrrolidinoethylamine

A121650-25G A121650-5G
ALA121650-25GEA 151.91 USD
ALA121650-25G ALA121650-5G
1-(2-Aminoethyl)pyrrolidine ≥98%
1-(2-Aminoethyl)pyrrolidine

1-(2-Aminoethyl)pyrrolidine is a heterocyclic building block. Nickel(II) salts of 1-(2-aminoethyl)pyrrolidine have been synthesized and thermal studies have been reported. The isomeric complexes trans-[NiL2(NCS)2] (violet) and cis-[NiL2(NCS)2] (blue) [L=1-(2-aminoethyl)pyrrolidine] have been synthesized and their X-ray crystal structure have been studied.Product Application:1-(2-Aminoethyl)pyrrolidine may be used in the preparation of one-dimensional coordination polymer of nickel(II), [(μN,S-NCS)2{Ni(ampy)}]n (ampy=1-(2-aminoethyl)pyrrolidine).

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 161°C
Flash Pt: 47°C
Storage Temperature: Ambient
MDL Number: MFCD00003182
CAS Number: 7154-73-6
EINECS: 230-509-5
UN: 2734
ADR: 3(8) II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60451948a08f28521f9e92efd8a041e3e031fa6c&vid=60451948a08f28521f9e92efd8a041e3e031fa6c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR