Ammonium chloride-[D4] ≥98 atom% D

Supplier: ALADDIN SCIENTIFIC

A121292-1G A121292-5G
ALA121292-1GEA 24.61 USD
ALA121292-1G ALA121292-5G
Ammonium chloride-[D4] ≥98 atom% D
Ammonium chloride-[D4]
Formula: ND₄Cl
Melting Pt: 340°C
Storage Temperature: Ambient
CAS Number: 12015-14-4
EINECS: 234-607-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR