3-Amino-2,6-dibromopyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2,6-Dibromopyridin-3-amine

A121064-25G A121064-5G A121064-1G
ALA121064-25GEA 483.46 USD
ALA121064-25G ALA121064-5G ALA121064-1G
3-Amino-2,6-dibromopyridine ≥98%
3-Amino-2,6-dibromopyridine
Formula: C₅H₄Br₂N₂
Melting Pt: 147°C
Storage Temperature: Ambient
MDL Number: MFCD00128862
CAS Number: 39856-57-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=044bfdb04eb377520b5407f475147bfa07dca6fa&vid=044bfdb04eb377520b5407f475147bfa07dca6fa&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR