(S)-(-)-2-Amino-2'-hydroxy-1,1'-binaphthyl ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
(S)-2'-Amino-[1,1'-binaphthalen]-2-ol, (S)-(-)-2'-Amino-1,1'-binaphthalen-2-ol, (S)-(-)-1-(2-Amino-1-naphthyl)-2-naphthol, (S)-(-)-NOBIN
A120831-100MG
A120831-500MG
ALA120831100MGEA
134.81
USD
ALA120831100MG
ALA120831500MG
(S)-(-)-2-Amino-2'-hydroxy-1,1'-binaphthyl ≥98%
(S)-(-)-2-Amino-2'-hydroxy-1,1'-binaphthyl
Formula:
C₂₀H₁₅NO Melting Pt: 173°C Storage Temperature: Ambient |
MDL Number:
MFCD01882346 CAS Number: 137848-29-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...