(R)-(+)-α,α-Diphenylprolinol ≥99%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: (R)-(+)-alpha,alpha-Diphenylprolinol, (R)-(+)-α,α-Diphenyl-2-pyrrolidinemethanol, (R)-(+)-2-(Diphenylhydroxymethyl)pyrrolidine, α,α-Diphenyl-D-prolinol

A120754-100G A120754-1G A120754-25G A120754-5G
ALA120754-100GEA 6813.31 USD
ALA120754-100G ALA120754-1G ALA120754-25G ALA120754-5G
(R)-(+)-α,α-Diphenylprolinol ≥99%
(R)-(+)-α,α-Diphenylprolinol

Used to prepare the corresponding oxazaborolidines for the borane-mediated asymmetric reduction of ketones.

Formula: C₁₇H₁₉NO
Melting Pt: 78°C
Storage Temperature: Ambient
MDL Number: MFCD00077754
CAS Number: 22348-32-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fbee3be09060c2cdbd3dea2d6e56b590b06fd23&vid=8fbee3be09060c2cdbd3dea2d6e56b590b06fd23&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR