4,5-Difluoroanthranilic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Amino-4,5-difluorobenzoic acid

A120663-100G A120663-1G A120663-25G A120663-5G
ALA120663-100GEA 549.01 USD
ALA120663-100G ALA120663-1G ALA120663-25G ALA120663-5G
4,5-Difluoroanthranilic acid ≥98%
4,5-Difluoroanthranilic acid
Formula: C₇H₅F₂NO₂
Melting Pt: 181-183°C
Storage Temperature: Ambient
MDL Number: MFCD00077509
CAS Number: 83506-93-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=496113168809704976d35403298358c08a7837a0&vid=496113168809704976d35403298358c08a7837a0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR