Ancymidol ≥98% (by HPLC) for plant cell culture

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: alpha-Cyclopropyl-alpha-(4-methoxyphenyl)-5-pyrimidinemethanol

A120564-100MG A120564-25MG
ALA120564100MGEA 499.61 USD
ALA120564100MG ALA120564-25MG
Ancymidol ≥98% (by HPLC) for plant cell culture
Ancymidol

Ancymidol is a pyrimidine-class plant cell endocrine disruptor plant growth regulator that interferes with gibberellin and cellulose biosynthesis.

Formula: C₁₅H₁₆N₂O₂
Density: 1.259 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00072501
CAS Number: 12771-68-5
EINECS: 235-814-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=943b50e12722ea3a8b2832bfc1a0c65d49c2e41d&vid=943b50e12722ea3a8b2832bfc1a0c65d49c2e41d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR