(S)-(+)-1-Boc-3-(aminomethyl)pyrrolidine ≥95%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
(S)-(+)-1-Boc-3-(aminomethyl)pyrrolidine, (S)-1-Boc-3-(Aminomethyl)pyrrolidine, (S)-3-(Aminomethyl)-1-(tert-butoxycarbonyl)pyrrolidine, (S)-3-(Aminomethyl)-1-Boc-pyrrolidine, tert-Butyl (S)-3-(aminomethyl)-1-pyrrolidinecarboxylate
A120235-5G
A120235-100MG
A120235-1G
A120235-25G
A120235-500MG
ALA120235-5GEA
1579.76
USD
ALA120235-5G
ALA120235100MG
ALA120235-1G
ALA120235-25G
ALA120235500MG
(S)-(+)-1-Boc-3-(aminomethyl)pyrrolidine ≥95%
(S)-(+)-1-Boc-3-(aminomethyl)pyrrolidine
Formula:
C₁₀H₂₀N₂O₂ MW: 200.282 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD02179397 CAS Number: 199175-10-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...