5-Aminoquinoline ≥97% (by titration)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
Quinolin-5-amine, Quinolin-5-ylamine
A120009-100G
A120009-1G
ALA120009-100GEA
299.16
USD
ALA120009-100G
ALA120009-1G
5-Aminoquinoline ≥97% (by titration)
5-Aminoquinoline
Formula:
C₉H₈N₂ MW: 144.18 g/mol Boiling Pt: 310 Melting Pt: 109°C Storage Temperature: Ambient |
MDL Number:
MFCD00006797 CAS Number: 611-34-7 EINECS: 210-266-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...