4-Amino-3,5-dichloropyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A119935-25G A119935-5G
ALA119935-25GEA 107.26 USD
ALA119935-25G ALA119935-5G
4-Amino-3,5-dichloropyridine ≥98%
4-Amino-3,5-dichloropyridine
Formula: C₅H₄Cl₂N₂
Melting Pt: 162°C
MDL Number: MFCD00125023
CAS Number: 22889-78-7
EINECS: 245-304-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc1c65b11ede0489f0ed2f442c9bb661311008e5&vid=bc1c65b11ede0489f0ed2f442c9bb661311008e5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR