4'-Aminobenzo-15-crown-5 (4'-aminobenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
2,3-(4-Aminobenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene, 4-Aminobenzo-15-crown 5-Ether, 4'-Aminobenzo-15-crown 5-Ether
A119932-1G
A119932-250MG
ALA119932-1GEA
159.51
USD
ALA119932-1G
ALA119932250MG
4'-Aminobenzo-15-crown-5 (4'-aminobenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥97%
4'-Aminobenzo-15-crown-5 (4'-Aminobenzo-1,4,7,10,13-pentaoxacyclopentadecane)
Formula:
C₁₄H₂₁NO₅ Melting Pt: 79°C Storage Temperature: Ambient |
MDL Number:
MFCD00068686 CAS Number: 60835-71-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...