3-Amino-4-methylbenzotrifluoride ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Methyl-5-(trifluoromethyl)aniline

A119814-100G A119814-1G A119814-25G A119814-5G
ALA119814-100GEA 2032.91 USD
ALA119814-100G ALA119814-1G ALA119814-25G ALA119814-5G
3-Amino-4-methylbenzotrifluoride ≥98%
3-Amino-4-methylbenzotrifluoride
Formula: C₈H₈F₃N
MW: 175.16 g/mol
Boiling Pt: 74 °C
Melting Pt: 36-40°C
MDL Number: MFCD00134580
CAS Number: 25449-96-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2a8947b48dfcfb7c5d04ae7af485dcd2d557840&vid=f2a8947b48dfcfb7c5d04ae7af485dcd2d557840&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR