3-Amino-2-bromo-5-fluoropyridine ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 2-Bromo-5-fluoropyridin-3-amine, 2-Bromo-5-fluoro-3-pyridylamine

A119665-1G A119665-250MG
ALA119665-1GEA 115.81 USD
ALA119665-1G ALA119665250MG
3-Amino-2-bromo-5-fluoropyridine ≥97%
3-Amino-2-bromo-5-fluoropyridine
Formula: C₅H₄BrFN₂
MW: 191
Melting Pt: 97-101°C
MDL Number: MFCD05662413
CAS Number: 884495-03-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42ee42a2bece87eb319caaf5879b7ba94151b19b&vid=42ee42a2bece87eb319caaf5879b7ba94151b19b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR