5-Aza-2'-deoxycytidine ≥98%, Moligand™
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
5-Aza-2′-deoxycytidine, Decitabine, 5-Aza-1-(2-deoxy-beta-D-ribofuranosyl)cytosine, 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,3,5-triazin-2(1H)-one, 2′-Deoxy-5-azacytidine 4-Amino-1-(2-deoxy-β-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one Decitabine, 4-Amino-1-(2-deoxy-beta-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one
Decitabine is an epigenetic modifier that inhibits DNA methyltransferase activity which results in DNA demethylation (hypomethylation) and gene activation by remodeling "opening" chromatin. Genes are synergistically reactivated when demethylation is combined with histone hyperacetylation.
Formula:
C₈H₁₂N₄O₄ Melting Pt: 191 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00043011 CAS Number: 2353-33-5 EINECS: 219-089-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...