Adenosine-5'-diphosphoric acid (ADP) ≥95% (by HPLC), Moligand™

Supplier: ALADDIN SCIENTIFIC

Synonyms: Adenosine 5'-diphosphate, 5′-ADP, Adenosine 5′-diphosphate

A119474-5G A119474-250MG A119474-25G A119474-1G
ALA119474-5GEA 245.96 USD
ALA119474-5G ALA119474250MG ALA119474-25G ALA119474-1G
Adenosine-5'-diphosphoric acid (ADP) ≥95% (by HPLC), Moligand™
Adenosine-5'-diphosphoric acid (ADP)
Formula: C₁₀H₁₅N₅O₁₀P₂
Storage Temperature: Ambient
MDL Number: MFCD00066473
CAS Number: 58-64-0
EINECS: 200-392-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f203f7b2d99e31810d674be84a590555e0804d8&vid=6f203f7b2d99e31810d674be84a590555e0804d8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR