Aluminium oxide nanoparticles 20% (w/w) in water 5-10 nm (TEM)
Supplier: ALADDIN SCIENTIFIC
A119404-100ML
A119404-25ML
ALA119404100MLEA
204.16
USD
ALA119404100ML
ALA119404-25ML
Aluminium oxide nanoparticles 20% (w/w) in water 5-10 nm (TEM)
Aluminium oxide
Formula:
Al₂O₃ Boiling Pt: 2977°C Melting Pt: 2000°C Density: 1.06 g/cm³ |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...