5-Aminouracil ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Aminouracil, 5-Aminopyrimidine-2,4(1H,3H)-dione

A119337-100G A119337-25G A119337-5G
ALA119337-100GEA 310.56 USD
ALA119337-100G ALA119337-25G ALA119337-5G
5-Aminouracil ≥98%
5-Aminouracil
Formula: C₄H₅N₃O₂
Melting Pt: >300°C
MDL Number: MFCD00006025
CAS Number: 932-52-5
EINECS: 213-252-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e1b285131af7cbc59fba74832ebde4514bf4226&vid=8e1b285131af7cbc59fba74832ebde4514bf4226&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR