2-Amino-4-chloropyrimidine ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
4-chloropyrimidin-2-amine
A119322-1G
A119322-25G
A119322-5G
ALA119322-1GEA
24.61
USD
ALA119322-1G
ALA119322-25G
ALA119322-5G
2-Amino-4-chloropyrimidine ≥98%
2-Amino-4-chloropyrimidine
Substrate used in a palladium-catalyzed cyanation with zinc(II) cyanide.
Formula:
C₄H₄ClN₃ Melting Pt: 155-160°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00038021 CAS Number: 3993-78-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...