Antimony(III) telluride ≥99.96% (metals basis)

Supplier: ALADDIN SCIENTIFIC
Danger

A119270-100G A119270-1G A119270-25G A119270-50G A119270-5G
ALA119270-100GEA 503.41 USD
ALA119270-100G ALA119270-1G ALA119270-25G ALA119270-50G ALA119270-5G
Antimony(III) telluride ≥99.96% (metals basis)
Antimony (III) telluride

Sb2Te3, along with Bi2Te3 and other structurally analogous semiconductors, is widely regarded as one of the best materials for room temperature thermoelectric devices. Antimony(III) telluride is a key component of thermoelectric materials that have reached ZT values as high as 2.4 at room temperature.

Formula: Sb₂Te₃
MW: 626.32 g/mol
Melting Pt: 620
MDL Number: MFCD00016320
CAS Number: 1327-50-0
UN: 1549
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR