(Acetonitrile)[(2-biphenyl)di-tert-butylphosphine]gold(I) hexafluoroantimonate 98

Supplier: ALADDIN SCIENTIFIC
Warning

A118699-1G A118699-250MG
ALA118699-1GEA 281.11 USD
ALA118699-1G ALA118699250MG
(Acetonitrile)[(2-biphenyl)di-tert-butylphosphine]gold(I) hexafluoroantimonate 98
(Acetonitrile)[(2-biphenyl)di-tert-butylphosphine]gold(I) hexafluoroantimonate
Formula: C₂₂H₃₀AuF₆NPSb
MW: 772.17 g/mol
Melting Pt: 198-203°C
Storage Temperature: Ambient
MDL Number: MFCD11045449
CAS Number: 866641-66-9
UN: 3077
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b8f39d8314c8fd6d2ca27349f63ff9b5aa9e64e&vid=4b8f39d8314c8fd6d2ca27349f63ff9b5aa9e64e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR