L(+)-Arginine ≥98.5%, Moligand™ USP, Ph. Eur. for cell culture animal-free

Supplier: ALADDIN SCIENTIFIC
Warning

A118651-100G A118651-25G
ALA118651-100GEA 94.91 USD
ALA118651-100G ALA118651-25G
L(+)-Arginine ≥98.5%, Moligand™ USP, Ph. Eur. for cell culture animal-free
L(+)-Arginine

L-Arginine is an essential amino acid and precursor of nitric oxide synthesis that induces vasodilation in vivo. Nitric oxide potentiates insulin-mediated glucose uptake through the increase in blood flow. A precursor of nitric oxide synthesis that induces vasodilationApplication:L-Arginine has been used:as a Roswell park memorial institute medium (RPMI) media component in the isolation and culture of peripheral blood mononuclear cells (PBMCs)as a RPMI media component for tissue culturein DMEM medium for the identification and quantification of phosphorylation sites by stable isotope labeling by amino acids in cell culture (SILAC) and LCMS/MS

Formula: C₆H₁₄N₄O₂
Melting Pt: 222 °C
Density: 1.2196 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00002635
CAS Number: 74-79-3
EINECS: 200-811-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e654b67384710d6c295151ede1fbf18d9dec50d&vid=0e654b67384710d6c295151ede1fbf18d9dec50d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR