Tributylallyltin 97

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: Allyltributylstannane, Allyltributyltin, Tributylallylstannane, Tributyl-2-propenylstannane, Allyltri-n-butyltin, Tri-n-butyl-2-propenylstannane, Allyltri-n-butylstannane

A118613-100G A118613-25G A118613-5G
ALA118613-100GEA 188.01 USD
ALA118613-100G ALA118613-25G ALA118613-5G
Tributylallyltin 97
Tributylallyltin

Aldehyde allylation reagent, the catalyst of this reaction is chiral Lewis acid and Ytterbium(III) trifluoromethanesulfonate. Catalyzed by tetrakis(triphenylphosphine)palladium (0) (product number 216666), it reacts with iodoquinone 4 and allyl acetate. Used in the efficient allylation reaction of aldehydes and trichloro-1,3,5-triazene to produce high allyl alcohol.

Formula: C₁₅H₃₂Sn
MW: 331.13 g/mol
Boiling Pt: 88
Flash Pt: 103 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010346
CAS Number: 24850-33-7
EINECS: 246-494-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8914e0d52fbe0c3acb7ffecd07a276b5f9917d1a&vid=8914e0d52fbe0c3acb7ffecd07a276b5f9917d1a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR