Acid red 27 ≥85%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Amaranth, Sodium 3-hydroxy-4-((4-sulfonatonaphthalen-1-yl)diazenyl)naphthalene-2,7-disulfonate, Sodium 3-hydroxy-4-((4-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-2,7-disulphonate, Bordeaux S, Food Red No. 2, Azo Rubine S, Wool Red 40 F, Naphthol Red

A109284-100G A109284-25G
ALA109284-100GEA 82.9 USD
ALA109284-100G ALA109284-25G
Acid red 27 ≥85%
Acid red 27
Formula: C₂₀H₁₄N₂O₁₀S₃
MW: 604.46 g/mol
Melting Pt: 218-220°C
Storage Temperature: Ambient
MDL Number: MFCD00004076
CAS Number: 915-67-3
EINECS: 213-022-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f24bdb3af51f48cad9d436da2ad7593018f612ca&vid=f24bdb3af51f48cad9d436da2ad7593018f612ca&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR