Adson Forceps, Serrated Tips
Supplier: WORLD PRECISION INSTRUMENTS LLC
14226 500222 500222-G 14226-G
77661-874EA 86.24 USD
77661-874 77662-190 77662-192 77661-876
Adson Forceps, Serrated Tips
Forceps
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.
  • Serrated
  • 1.5 mm tips