ESR Pipette
Supplier: SARSTEDT INC
86.1996 86.1996.062
77589-596CS 1108.98 USD
77589-596 77589-598
ESR Pipette
Pipette Tips
Full suite of solutions for ESR testing products with the pipettes, Sediplus®, S-Sedivette®, and Microvette® CB 200 ESR.
  • Automatic and manual determination in accordance with Westergren
  • Simple adjustment of blood column to zero through thread
  • Bubble free filling of pipettes
The Sediplus® S 2000 NX is the solution to automatic blood sedimentation analysis. It is operated with a touch display and intuitive menu navigation. In addition, a printer and scanner can be added to the device.
The S-Sedivette® allows proven closed venous blood collection with choice of aspiration or vacuum technique. 2.8 ml blood are added to the S-Sedivette® pre-dosed with citrate (mixing ratio 1:4).
Packaging: The ESR measurement is carried out directly in the S-Sedivette® as a closed system.