Cell Culture Dishes, lumox® dish
Supplier: SARSTEDT INC
94.6077.305 94.6077.331 94.6077.333 94.6077.410
77589-828CS 3200.91 USD
77589-828 77589-832 77589-834 77589-836
Cell Culture Dishes, lumox® dish
Culture Dishes Cell Culture Dishes
To keep cells happy, the selection of a suitable cultivation surface is essential. SARSTEDT offers a wide range of cell culture labware and accessories for many applications.
  • Material: lumox®
  • Frame material: PS
  • Purity: Sterile, non-pyrogenic/endotoxin-free, non-cytotoxic
  • Cell culture dishes with lumox® film base
  • Low autofluorescence and higher light transmission than polystyrene or glass bases
  • Gas-permeable film base guarantees optimal gas exchange
  • Cultivation flasks for adherent cells or suspension cells
  • Range of applications from normal cell culture to the automated analysis of fluorescence-based cell assays
The thin, gas-permeable film base of lumox® cell products ensures optimal cell growth through short diffusion paths and excellent gas exchange. lumox® film bases also have excellent optical properties. Minimal autofluorescence and high light transmission result in a high sensitivity with fluorescence-based assays. SARSTEDT offers lumox® dishes and plates as well as x-well cell culture chambers for direct cultivation and analysis of cells on a slide.