Accessories for miniPERM® Bioreactors
Supplier: SARSTEDT INC
94.6077.037 94.6077.138 94.6001.036 94.6001.054 94.6001.153
77589-822CS 655.39 USD
77589-822 77589-826 77589-798 77589-800 77589-806
Accessories for miniPERM® Bioreactors
Bioreactors
To keep cells happy, the selection of a suitable cultivation surface is essential. SARSTEDT offers a wide range of cell culture labware and accessories for many applications.
  • Accessories for the easy handling of the miniPERM® bioreactor
  • Autoclavable supply module for use with the sterile disposable production modules
  • Rack to place the miniPERM® bioreactor overhead under the sterile bench, ensures easy access to the production module
  • Caps, syringes and other consumables
  • Accessories are available individually or as a start-up support kit (includes all accessories that are required for initially starting the culture, sampling and harvesting)
The SARSTEDT miniPERM® is an easy-to-use bioreactor for the cultivation of eukaryotic suspension cells at a high density with the aim of producing proteins, viruses or biomass. The miniPERM® bioreactor consists of a production and a nutrient module connected by a dialysis membrane. It enables the production of highly concentrated cell products in the small volume of the production module, while supplying the cells with nutrients from the nutrient module. The gas is exchanged through a gas-permeable silicone membrane on the outside of the production module.