Wire Speculums, 4 cm
Supplier: WORLD PRECISION INSTRUMENTS LLC
501993 501994
77663-012EA 49.3 USD
77663-012 77663-014
Wire Speculums, 4 cm
Specula
A wire eyelid speculum is a surgical instrument used by ophthalmologists to keep eyelids open during an exam or surgery.
  • Small, wire eyelid speculum
  • Maximum spread: 15 mm
  • Surgical grade stainless steel
  • Autoclavable
It consists of two curved blades that are gently inserted between the upper and lower eyelids to hold them open, allowing the ophthalmologist a clear view of the eye. Our 4 cm wire specula are made of surgical grade stainless steel, and they come in two sizes, with a spread of 15 mm. This wire speculum can be sterilised with a chemical sterilant or by autoclaving.
Note: Be sure to clean, disinfect and sterilise surgical instruments promptly after each use to ensure they last for many years.