Ethyl 2-(2-fluoro-3-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%
Supplier: AOBChem USA
|
Warning
|
94682-5G
94682-25G
94682-1G
94682-500MG
77630-920EA
1441
USD
77630-920
77636-266
77640-240
77652-234
Ethyl 2-(2-fluoro-3-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(2-fluoro-3-(trifluoromethyl)phenyl)-2-oxoacetate
Caution: For Research Use Only
Formula:
C₁₁H₈F₄O₃ Storage Temperature: Ambient |
MDL Number:
MFCD26686630 CAS Number: 1508871-47-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...