1-(2-Chloro-4-fluorophenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

93160-5G 93160-25G 93160-1G 93160-500MG
77630-876EA 1441 USD
77630-876 77660-552 77648-192 77648-194
1-(2-Chloro-4-fluorophenyl)cyclobutanol ≥95%
1-(2-Chloro-4-fluorophenyl)cyclobutanol

Caution: For Research Use Only

Formula: C₁₀H₁₀ClFO
Storage Temperature: Ambient
MDL Number: MFCD34773436
CAS Number: 2918916-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR