1,3-Diethyl-5-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

92200-1G 92200-5G 92200-500MG
77640-186EA 580 USD
77640-186 77660-540 77652-196
1,3-Diethyl-5-iodobenzene ≥95%
1,3-Diethyl-5-iodobenzene

Caution: For Research Use Only

Formula: C₁₀H₁₃I
Storage Temperature: Ambient
MDL Number: MFCD29096239
CAS Number: 1369882-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR