Ethyl 2-(6-bromo-2,3-difluorophenyl)-2-oxoacetate ≥95%
Supplier: AOBChem USA
|
Warning
|
82679-1G
82679-5G
82679-500MG
77640-042EA
531
USD
77640-042
77656-944
77652-058
Ethyl 2-(6-bromo-2,3-difluorophenyl)-2-oxoacetate ≥95%
Ethyl 2-(6-bromo-2,3-difluorophenyl)-2-oxoacetate
Caution: For Research Use Only
Formula:
C₁₀H₇BrF₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD34769322 CAS Number: 1252799-90-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...