3-(2-Bromopyridin-4-yl)-1,1,1-trifluoropropan-2-one ≥97%

Supplier: AOBChem USA

Warning

76957-5G 76957-25G 76957-1G 76957-500MG
77630-666EA 1441 USD
77630-666 77656-882 77644-006 77652-000
3-(2-Bromopyridin-4-yl)-1,1,1-trifluoropropan-2-one ≥97%
3-(2-Bromopyridin-4-yl)-1,1,1-trifluoropropan-2-one

Caution: For Research Use Only

Formula: C₈H₅BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD34766763
CAS Number: 2901098-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR