5-(Dimethylamino)nicotinic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
76691-5G
76691-1G
76691-25G
76691-500MG
77633-710EA
1212
USD
77633-710
77639-986
77656-880
77651-998
5-(Dimethylamino)nicotinic acid ≥97%
5-(Dimethylamino)nicotinic acid
Caution: For Research Use Only
Formula:
C₈H₁₀N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD04971940 CAS Number: 30766-20-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...