1-(3-Chloro-5-(methylthio)phenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

64400-5G 64400-1G 64400-25G 64400-500MG
77630-572EA 1441 USD
77630-572 77639-878 77656-770 77647-862
1-(3-Chloro-5-(methylthio)phenyl)-2,2,2-trifluoroethanone ≥97%
1-(3-Chloro-5-(methylthio)phenyl)-2,2,2-trifluoroethanone

Caution: For Research Use Only

Formula: C₉H₆ClF₃OS
Storage Temperature: Ambient
MDL Number: MFCD34761752
CAS Number: 2918952-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR