2,4-Difluoro-5-iodo-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62443-5G 62443-25G 62443-1G 62443-500MG
77635-252EA 466 USD
77635-252 77656-722 77643-872 77651-848
2,4-Difluoro-5-iodo-3-methoxybenzaldehyde ≥97%
2,4-Difluoro-5-iodo-3-methoxybenzaldehyde

Caution: For Research Use Only

Formula: C₈H₅F₂IO₂
Storage Temperature: Ambient
MDL Number: MFCD34760841
CAS Number: 2764733-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR