1-(3-Bromo-5-methylphenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

62340-5G 62340-25G 62340-1G 62340-500MG
77630-546EA 1488 USD
77630-546 77636-186 77643-868 77651-844
1-(3-Bromo-5-methylphenyl)-2,2,2-trifluoroethanone ≥97%
1-(3-Bromo-5-methylphenyl)-2,2,2-trifluoroethanone

Caution: For Research Use Only

Formula: C₉H₆BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD28059739
CAS Number: 1823889-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR