2-(Benzo[b]thiophen-2-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

554979-5G 554979-25G 554979-1G 554979-500MG
77633-566EA 1171 USD
77633-566 77660-042 77643-704 77651-640
2-(Benzo[b]thiophen-2-yl)propan-2-ol ≥95%
2-(Benzo[b]thiophen-2-yl)propan-2-ol

Caution: For Research Use Only

Formula: C₁₁H₁₂OS
Storage Temperature: Ambient
CAS Number: 135345-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR