2,2,2-Trifluoro-1-(7-methylbenzo[b]thiophen-2-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

519038-5G 519038-1G 519038-25G 519038-500MG
77630-196EA 1484.23 USD
77630-196 77639-520 77656-390 77651-500
2,2,2-Trifluoro-1-(7-methylbenzo[b]thiophen-2-yl)ethanone ≥95%
2,2,2-Trifluoro-1-(7-methylbenzo[b]thiophen-2-yl)ethanone

Caution: For Research Use Only

Formula: C₁₁H₇F₃OS
Storage Temperature: Ambient
CAS Number: 3037703-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR