1-(5-Cyclohexyl-2-fluorophenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

487470-5G 487470-1G 487470-25G 487470-500MG
77629-910EA 1484.23 USD
77629-910 77639-290 77659-742 77651-280
1-(5-Cyclohexyl-2-fluorophenyl)ethanol ≥95%
1-(5-Cyclohexyl-2-fluorophenyl)ethanol

Caution: For Research Use Only

Formula: C₁₄H₁₉FO
Storage Temperature: Ambient
CAS Number: 3037703-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR