4-Methyl-5-thiazolecarbonitrile ≥97%
Supplier: AOBChem USA
|
Warning
|
468596-1G
468596-5G
468596-500MG
77639-050EA
615
USD
77639-050
77659-544
77651-048
4-Methyl-5-thiazolecarbonitrile ≥97%
4-Methyl-5-thiazolecarbonitrile
Caution: For Research Use Only
Formula:
C₅H₄N₂S Storage Temperature: Ambient |
CAS Number:
408526-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...