(2-Fluoro-3-propoxyphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

465057-5G 465057-25G 465057-1G 465057-500MG
77633-246EA 1441 USD
77633-246 77655-924 77643-002 77646-976
(2-Fluoro-3-propoxyphenyl)trimethylsilane ≥95%
(2-Fluoro-3-propoxyphenyl)trimethylsilane

Caution: For Research Use Only

Formula: C₁₂H₁₉FOSi
Storage Temperature: Ambient
CAS Number: 3037703-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR