2-(2-Bromo-6-fluorophenyl)propan-2-ol ≥97%
Supplier: AOBChem USA
|
Warning
|
448871-1G
448871-500MG
448871-5G
77638-828EA
558
USD
77638-828
77652-828
77655-784
2-(2-Bromo-6-fluorophenyl)propan-2-ol ≥97%
2-(2-Bromo-6-fluorophenyl)propan-2-ol
Caution: For Research Use Only
Formula:
C₉H₁₀BrFO Storage Temperature: Ambient |
CAS Number:
1514060-98-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...