Goldstein Retractors
Supplier: WORLD PRECISION INSTRUMENTS LLC
501967 501968 501969
77662-962EA 266.41 USD
77662-962 77662-964 77662-966
Goldstein Retractors
Retractors
Goldstein Retractors are surgical instrument used mostly for plastic and reconstructive surgery. Retractors expose and hold back tissues, giving you optimal visibility and access to the surgical site.
  • 3×3 sharp prongs
  • Small, self-retaining retractor perfect for small animal surgery and dissection
  • Sizes range from 2 to 4 cm long with a maximum spread between 1.0 and 3.5 cm
  • Surgical grade stainless steel
The Goldstein retractor is used for surgeries where precise tissue manipulation is required. For small animal surgery, a Goldstein retractor can be used to gently retract the skin and soft tissues at the incision site, allowing you to access the underlying anatomical structures. In procedures involving blood vessels, the Goldstein retractor can help expose and isolate vessels for vascular anastomosis or other manipulations. In some neurosurgical procedures on small animals, the Goldstein retractor may be used to retract the scalp and soft tissues, providing access to the skull and brain.
The Goldstein retractor is designed to be atraumatic, minimizing damage to tissues, which is especially important in laboratory settings. WPI's Goldstein Retractors have 3 sharp prongs on each blade and a knurled thumbscrews to adjust the spread of the blades and lock them into place. They come in multiples sizes with a maximum spread between 1 and 3.5 cm. These are manufactured from surgical grade stainless steel and are suitable for autoclaving or chemical sterilization. Clean surgical instruments promptly after use to ensure a long life for your surgical tools.